COMH93D665D3 Combi-Blocks, Inc.

3-Methyl-1-nonyn-3-ol

  • MDL number MFCD00041582
  • Cas Number : 5430-01-3
  • Molecular Weight : 154.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3847

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3847
Availability details for
Quantity
  • Availability
  • Add to Cart