COMH93D665EB Combi-Blocks, Inc.

2-Bromo-4,5-dimethoxycinnamic acid

  • MDL number MFCD00017614
  • Cas Number : 195383-80-3
  • Molecular Weight : 287.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3941

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3941
Availability details for
Quantity
  • Availability
  • Add to Cart