COMH93D666EE Combi-Blocks, Inc.

Calcium phosphate, monobasic, monohydrate

  • MDL number MFCD00149602
  • Cas Number : 301524-28-7
  • Molecular Weight : 252.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4873
Availability details for
Quantity
  • Availability
  • Add to Cart