COMH93D667A2 Combi-Blocks, Inc.

Cresyl diphenyl phosphate

  • MDL number MFCD00014920
  • Cas Number : 26444-49-5
  • Molecular Weight : 340.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methylphenyl diphenyl phosphate
Inchi Code 1S/C19H17O4P/c1-16-9-8-14-19(15-16)23-24(20,21-17-10-4-2-5-11-17)22-18-12-6-3-7-13-18/h2-15H,1H3
Inchi Key FJKVRVFGUXMMAO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5500
Availability details for
Quantity
  • Availability
  • Add to Cart