COMH93D66806 Combi-Blocks, Inc.

Acetaldehyde di-n-hexyl acetal

  • MDL number MFCD00051715
  • Cas Number : 5405-58-3
  • Molecular Weight : 230.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5832
Availability details for
Quantity
  • Availability
  • Add to Cart