COMH93D6681D Combi-Blocks, Inc.

N-Benzoylsuccinimide

  • MDL number MFCD00185021
  • Cas Number : 6343-27-7
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5901

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5901
Availability details for
Quantity
  • Availability
  • Add to Cart