COMH93D66835 Combi-Blocks, Inc.

Neodymium(iii) chloride hydrate

  • MDL number MFCD00149800
  • Cas Number : 10024-93-8
  • Molecular Weight : 268.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5987

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5987
Availability details for
Quantity
  • Availability
  • Add to Cart