COMH93D6684B Combi-Blocks, Inc.

1-(2-Furyl)butan-1-one

  • MDL number MFCD00046111
  • Cas Number : 4208-57-5
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6054

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6054
Availability details for
Quantity
  • Availability
  • Add to Cart