COMH93D668CC Combi-Blocks, Inc.

Dimethallyl carbonate

  • MDL number MFCD00008598
  • Cas Number : 64057-79-0
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6514
Availability details for
Quantity
  • Availability
  • Add to Cart