COMH93D66926 Combi-Blocks, Inc.

N-Methyldiacetamide

  • MDL number MFCD00014968
  • Cas Number : 1113-68-4
  • Molecular Weight : 115.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6815

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6815
Availability details for
Quantity
  • Availability
  • Add to Cart