COMH93D66937 Combi-Blocks, Inc.

1-Octadecanesulfonyl chloride

  • MDL number MFCD00051521
  • Cas Number : 10147-41-8
  • Molecular Weight : 353.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6870

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6870
Availability details for
Quantity
  • Availability
  • Add to Cart