COMH93D6693A Combi-Blocks, Inc.

(S)-(+)-1-(3-Methoxyphenyl)ethyl isothiocyanate

  • MDL number MFCD05664056
  • Cas Number : 737000-99-6
  • Molecular Weight : 193.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6874

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6874
Availability details for
Quantity
  • Availability
  • Add to Cart