COMH93D6696B Combi-Blocks, Inc.

6,7,8,9,10,11-Hexahydro-5h-cycloocta[b]indole

  • MDL number MFCD00086346
  • Cas Number : 22793-63-1
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7040

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7040
Availability details for
Quantity
  • Availability
  • Add to Cart