COMH93D6696C Combi-Blocks, Inc.

Ethyl 4-(1-methylethyl)piperazine-1-carboxylate

  • MDL number MFCD08064222
  • Cas Number : 61014-91-3
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-isopropyl-1-piperazinecarboxylate
Inchi Code 1S/C10H20N2O2/c1-4-14-10(13)12-7-5-11(6-8-12)9(2)3/h9H,4-8H2,1-3H3
Inchi Key HRMZRPQIFGCXML-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7045
Availability details for
Quantity
  • Availability
  • Add to Cart