COMH93D66992 Combi-Blocks, Inc.

Methylcyclopentadiene dimer

  • MDL number MFCD00082297
  • Cas Number : 26472-00-4
  • Molecular Weight : 160.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3aR,4S,7R,7aS)-2,5-dimethyl-3a,4,7,7a-tetrahydro-1H-4,7-methanoindene
Inchi Code 1S/C12H16/c1-7-3-11-9-5-8(2)10(6-9)12(11)4-7/h4-5,9-12H,3,6H2,1-2H3/t9-,10-,11?,12?/m1/s1
Inchi Key IYQYZZHQSZMZIG-QYNFOATHSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7171

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7171
Availability details for
Quantity
  • Availability
  • Add to Cart