COMH93D669A9 Combi-Blocks, Inc.

1,3-Diphenoxy-2-propanol

  • MDL number MFCD00020661
  • Cas Number : 622-04-8
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7244

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7244
Availability details for
Quantity
  • Availability
  • Add to Cart