COMH93D669B2 Combi-Blocks, Inc.

3-Chloro-5-(pentafluorosulfur)benzaldehyde

  • MDL number MFCD16652412
  • Cas Number : 1240257-88-8
  • Molecular Weight : 266.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7266
Availability details for
Quantity
  • Availability
  • Add to Cart