COMH93D669B7 Combi-Blocks, Inc.

3-Methylbenzoic anhydride

  • MDL number MFCD00048083
  • Cas Number : 21436-44-2
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7279

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7279
Availability details for
Quantity
  • Availability
  • Add to Cart