COMH93D669C3 Combi-Blocks, Inc.

4-(1,3-Dioxolan-2-yl)benzonitrile

  • MDL number MFCD00052570
  • Cas Number : 66739-89-7
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7327
Availability details for
Quantity
  • Availability
  • Add to Cart