COMH93D669CB Combi-Blocks, Inc.

Methyl 2,1,3-benzothiadiazole-5-carboxylate

  • MDL number MFCD00173948
  • Cas Number : 175204-21-4
  • Molecular Weight : 194.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2,1,3-benzothiadiazole-5-carboxylate
Inchi Code 1S/C8H6N2O2S/c1-12-8(11)5-2-3-6-7(4-5)10-13-9-6/h2-4H,1H3
Inchi Key LRMFSMKHXKDUDG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7347
Availability details for
Quantity
  • Availability
  • Add to Cart