COMH93D669CD Combi-Blocks, Inc.

2,4-Dimethyl-2-pentanol

  • MDL number MFCD00021811
  • Cas Number : 625-06-9
  • Molecular Weight : 116.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-dimethyl-2-pentanol
Inchi Code 1S/C7H16O/c1-6(2)5-7(3,4)8/h6,8H,5H2,1-4H3
Inchi Key FMLSQAUAAGVTJO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7350

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7350
Availability details for
Quantity
  • Availability
  • Add to Cart