COMH93D66A1B Combi-Blocks, Inc.

Dimethyl allylphosphonate

  • MDL number MFCD00014951
  • Cas Number : 757-54-0
  • Molecular Weight : 150.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7643

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7643
Availability details for
Quantity
  • Availability
  • Add to Cart