COMH93D66C2E Combi-Blocks, Inc.

4-Dodecanone

  • MDL number MFCD00015300
  • Cas Number : 6137-26-4
  • Molecular Weight : 184.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9492

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9492
Availability details for
Quantity
  • Availability
  • Add to Cart