COMH93D66C88 Combi-Blocks, Inc.

(2-Chloroethyl)triphenylphosphonium bromide

  • MDL number MFCD00051877
  • Cas Number : 31238-20-7
  • Molecular Weight : 405.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9833

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9833
Availability details for
Quantity
  • Availability
  • Add to Cart