COMH93D66C8C Combi-Blocks, Inc.

Diethyl 1-octylphosphonate

  • MDL number MFCD00015659
  • Cas Number : 1068-07-1
  • Molecular Weight : 250.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9855

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9855
Availability details for
Quantity
  • Availability
  • Add to Cart