COMH93D66CBC Combi-Blocks, Inc.

2-Fluoro-4-(pentafluorosulfur)benzamide

  • MDL number MFCD16652504
  • Cas Number : 1240257-30-0
  • Molecular Weight : 265.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0036
Availability details for
Quantity
  • Availability
  • Add to Cart