COMH93D66CC3 Combi-Blocks, Inc.

(R)-(+)-1-(4-Fluorophenyl)ethyl isocyanate

  • MDL number MFCD05664075
  • Cas Number : 745783-73-7
  • Molecular Weight : 165.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0067
Availability details for
Quantity
  • Availability
  • Add to Cart