COMH93D66CCA Combi-Blocks, Inc.

2-Chloro-3,6-difluoroaniline

  • MDL number MFCD06660176
  • Cas Number : 287172-80-9
  • Molecular Weight : 163.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0104

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0104
Availability details for
Quantity
  • Availability
  • Add to Cart