COMH93D66CD0 Combi-Blocks, Inc.

4-(3-Methoxyphenoxy)piperidine

  • MDL number MFCD08061092
  • Cas Number : 162402-37-1
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0142

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0142
Availability details for
Quantity
  • Availability
  • Add to Cart