COMH93D66CD6 Combi-Blocks, Inc.

4-(4-Bromophenyl)-2-methylpyrimidine

  • MDL number MFCD02089754
  • Cas Number : 499785-50-1
  • Molecular Weight : 249.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0170

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0170
Availability details for
Quantity
  • Availability
  • Add to Cart