COMH93D66CDC Combi-Blocks, Inc.

Alpha-(4-tert-butylphenyl)di(2-pyrrolyl)methane

  • MDL number MFCD09027316
  • Cas Number : 167482-98-6
  • Molecular Weight : 278.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0203

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0203
Availability details for
Quantity
  • Availability
  • Add to Cart