COMH93D66D0F Combi-Blocks, Inc.

4-Methoxy-3-(trifluoromethoxy)phenylacetic acid

  • MDL number MFCD18398521
  • Cas Number : 1261557-46-3
  • Molecular Weight : 250.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-methoxy-3-(trifluoromethoxy)phenyl]acetic acid
Inchi Code 1S/C10H9F3O4/c1-16-7-3-2-6(5-9(14)15)4-8(7)17-10(11,12)13/h2-4H,5H2,1H3,(H,14,15)
Inchi Key MMYRFHYSHNVBCU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0463
Availability details for
Quantity
  • Availability
  • Add to Cart