COMH93D66D21 Combi-Blocks, Inc.

5-Chloro-2-(trifluoromethoxy)phenylacetic acid

  • MDL number MFCD11519360
  • Cas Number : 1092461-21-6
  • Molecular Weight : 254.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0612
Availability details for
Quantity
  • Availability
  • Add to Cart