COMH93D66D45 Combi-Blocks, Inc.

2-Methoxy-5-(trifluoromethyl)cinnamic acid

  • MDL number MFCD06205858
  • Cas Number : 773133-16-7
  • Molecular Weight : 246.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0914

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0914
Availability details for
Quantity
  • Availability
  • Add to Cart