COMH93D66D47 Combi-Blocks, Inc.

N-Ethyl-2-methoxy-n-(3-methylphenyl)benzamide

  • MDL number MFCD00086720
  • Cas Number : 199106-97-3
  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0923

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0923
Availability details for
Quantity
  • Availability
  • Add to Cart