COMH93D66D56 Combi-Blocks, Inc.

1-(4-Piperidinyloxy)acetylpyrrolidine

  • MDL number MFCD08061028
  • Cas Number : 902836-88-8
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[2-oxo-2-(1-pyrrolidinyl)ethoxy]piperidine
Inchi Code 1S/C11H20N2O2/c14-11(13-7-1-2-8-13)9-15-10-3-5-12-6-4-10/h10,12H,1-9H2
Inchi Key TZJOTOFHDQDADH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1032

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1032
Availability details for
Quantity
  • Availability
  • Add to Cart