COMH93D66D64 Combi-Blocks, Inc.

N-(4-Bromo-2-fluorophenyl)-4-chlorobenzamide

  • MDL number MFCD01359597
  • Cas Number : 519017-62-0
  • Molecular Weight : 328.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1105
Availability details for
Quantity
  • Availability
  • Add to Cart