COMH93D66D85 Combi-Blocks, Inc.

(4S,5S)-(+)-4,5-Bis(fluoromethyl)-2,2-dimethyl-1,3-dioxolane

  • MDL number MFCD01074597
  • Cas Number : 84409-70-1
  • Molecular Weight : 166.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4S,5S)-4,5-bis(fluoromethyl)-2,2-dimethyl-1,3-dioxolane
Inchi Code 1S/C7H12F2O2/c1-7(2)10-5(3-8)6(4-9)11-7/h5-6H,3-4H2,1-2H3/t5-,6-/m1/s1
Inchi Key LFHJRQWZIPGXAM-PHDIDXHHSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1292
Availability details for
Quantity
  • Availability
  • Add to Cart