COMH93D66D97 Combi-Blocks, Inc.

3,4,6-Tri-o-methyl-d-glucal

  • MDL number MFCD22989010
  • Cas Number : 16740-98-0
  • Molecular Weight : 188.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1405

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1405
Availability details for
Quantity
  • Availability
  • Add to Cart