COMH93D66DAA Combi-Blocks, Inc.

1-(4-Methylphenyl)homopiperazine monohydrochloride

  • MDL number MFCD16172201
  • Cas Number : 1192191-39-1
  • Molecular Weight : 226.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-methylphenyl)-1,4-diazepane hydrochloride
Inchi Code 1S/C12H18N2.ClH/c1-11-3-5-12(6-4-11)14-9-2-7-13-8-10-14;/h3-6,13H,2,7-10H2,1H3;1H
Inchi Key WOAJKNSSUFZJHK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1572
Availability details for
Quantity
  • Availability
  • Add to Cart