COMH93D66DB7 Combi-Blocks, Inc.

2-Chloro-n-ethyl-n-(3-methylphenyl)benzamide

  • MDL number MFCD22683085
  • Cas Number : 1018134-12-7
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-N-ethyl-N-m-tolylbenzamide
Inchi Code 1S/C16H16ClNO/c1-3-18(13-8-6-7-12(2)11-13)16(19)14-9-4-5-10-15(14)17/h4-11H,3H2,1-2H3
Inchi Key YFBNVKAYRHEGFA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1661
Availability details for
Quantity
  • Availability
  • Add to Cart