COMH93D66DE8 Combi-Blocks, Inc.

2-(N-Propylthio)nicotinamide

  • MDL number MFCD00052904
  • Cas Number : 175135-26-9
  • Molecular Weight : 196.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(propylsulfanyl)nicotinamide
Inchi Code 1S/C9H12N2OS/c1-2-6-13-9-7(8(10)12)4-3-5-11-9/h3-5H,2,6H2,1H3,(H2,10,12)
Inchi Key JIYBJKGFAMWNDO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1964
Availability details for
Quantity
  • Availability
  • Add to Cart