COMH93D66E34 Combi-Blocks, Inc.

4-Chloro-2-fluoro-3-methoxy-dl-phenylalanine

  • MDL number MFCD19687211
  • Cas Number : 1706418-83-8
  • Molecular Weight : 247.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2373

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2373
Availability details for
Quantity
  • Availability
  • Add to Cart