COMH93D66E49 Combi-Blocks, Inc.

1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1h-pyrazole-4-sulfonyl chloride

  • MDL number MFCD11505058
  • Cas Number : 1215564-15-0
  • Molecular Weight : 312.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2547

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2547
Availability details for
Quantity
  • Availability
  • Add to Cart