COMH93D6703F Combi-Blocks, Inc.

2-Amino-5-iodo-4,6-dimethylpyridine

  • MDL number MFCD06798226
  • Cas Number : 885952-12-5
  • Molecular Weight : 248.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5664

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5664
Availability details for
Quantity
  • Availability
  • Add to Cart