COMH93D6704B Combi-Blocks, Inc.

2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetonitrile

  • MDL number MFCD22201045
  • Cas Number : 1373920-64-9
  • Molecular Weight : 233.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5760

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5760
Availability details for
Quantity
  • Availability
  • Add to Cart