COMH93D6704E Combi-Blocks, Inc.

2-Fluoro-5-(pentafluorosulfur)aniline

  • MDL number MFCD16652446
  • Cas Number : 1240257-94-6
  • Molecular Weight : 237.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5788

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5788
Availability details for
Quantity
  • Availability
  • Add to Cart