COMH93D6706D Combi-Blocks, Inc.

3-(2-Methyl-3-furylthio)-4-heptanone

  • MDL number MFCD01723735
  • Cas Number : 61295-41-8
  • Molecular Weight : 226.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6005
Availability details for
Quantity
  • Availability
  • Add to Cart