COMH93D67074 Combi-Blocks, Inc.

2-Methyl-5-(trifluoromethoxy)phenylacetonitrile

  • MDL number MFCD18399826
  • Cas Number : 1261683-78-6
  • Molecular Weight : 215.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6074

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6074
Availability details for
Quantity
  • Availability
  • Add to Cart