COMH93D6707F Combi-Blocks, Inc.

3-(3-Oxocyclopentyl)indole-5-carbonitrile

  • MDL number MFCD28411341
  • Cas Number : 675831-64-8
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6163
Availability details for
Quantity
  • Availability
  • Add to Cart